L(+)-Histidin Hydrochlorid Monohydrat, Millipore®

Lieferant: Merck

Synonyme: His HCl Monohydrat, (S)-(+)-Histidin Hydrochlorid Monohydrat, (2S)-(+)-Histidin Hydrochlorid Monohydrat, (S)-(+)-2-Amino-3-(4-imidazoyl)propansäure Hydrochlorid Monohydrat, L-3-Imidazol-4-ylalanin Hydrochlorid Monohydrat, H-His-OH.HCl.H2O

M-Clarity™ Qualitätsstufe = MQ100

1.04350.0025 1.04350.0100 1.04350.0500 1.04350.9010
1.04350.0025EA 62.1 EUR
1.04350.0025 1.04350.0100 1.04350.0500 1.04350.9010
L(+)-Histidin Hydrochlorid Monohydrat, Millipore®
L(+)-Histidin Hydrochlorid Monohydrat
Formel: C₆H₉N₃O₂·HCl·H₂O
Schmelzpunkt: 259 °C
Dichte: 1,485 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00151027
CAS-Nummer: 5934-29-2
EINECS: 211-438-9
Merck Index: 13,04741
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=561d2381f930afe618f78d680790ea9a43f6596c&vid=8aacf6dc4ef5382410cb743498272b48c3c36999&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR