L(+)-Histidin, Millipore®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

1.04351.0100 1.04351.1000 1.04351.9010
1.04351.0100EA 124 EUR
1.04351.0100 1.04351.1000 1.04351.9010
L(+)-Histidin, Millipore®
L(+)-Histidin

Soluble in water and mineral acids; insoluble in organic solvents.

Formel: C₆H₉N₃O₂
Schmelzpunkt: 272…273 °C
Dichte: 1,44 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064315
CAS-Nummer: 71-00-1
EINECS: 200-745-3
Merck Index: 13,04741
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=badfdb36aac0611e5823a7d05e6cba944f6650fa&vid=9e9f9d881a5b3314c7f342312e77e13493901200&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR