Quecksilber(II)nitrat Monohydrat ACS, Reag. Ph. Eur. zur Analyse, Supelco®
Lieferant: Merck
1.04439.0050
1.04439.0250
1.04439.0050EA
102
EUR
1.04439.0050
1.04439.0250
Quecksilber(II)nitrat Monohydrat ACS, Reag. Ph. Eur. zur Analyse, Supelco®
Spezifikation Testergebnisse
Assay (complexometric) | ≥ 99.0 % |
Chloride (Cl) | ≤ 0.002 % |
Sulfate (SO₄) | ≤ 0.002 % |
Cu (Copper) | ≤ 0.0005 % |
Fe (Iron) | ≤ 0.001 % |
K (Potassium) | ≤ 0.005 % |
Na (Sodium) | ≤ 0.005 % |
Residue after reduction | ≤ 0.01 % |
Corresponds to ACS,Reag. Ph Eur |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...