Benzonase® Salt Tolerant Endonuclease
Lieferant: Merck
The best-in-class solution for nucleic acid removal at high salt concentration in Bioprocessing. The Benzonase® Salt Tolerant endonuclease Emprove® Expert is a new enzyme, different in amino acids sequence from other Benzonase® endonuclease references. It was designed with state-of-the-art protein engineering capabilities, to ensure the highest activity at high salt concentrations.
- Application: Pharma/biopharma processes
The protein is a monomer with a molecular weight of about 27 kDa . The protein has an isoelectric point (pI) at pH 9,68. It is functional between pH 6 and 10, and from 0 °C to above 37 °C. Mg²⁺ (1 to 2 mM) is required for enzyme activity.