Quecksilber(II)oxid, rot zur Analyse, Supelco®

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ300

1.04466.0050 1.04466.0250
1.04466.0050EA 125 EUR
1.04466.0050 1.04466.0250
Quecksilber(II)oxid, rot zur Analyse, Supelco®
Mercury (II) oxide
Formel: HgO
Molecular Weight: 216.59 g/mol
Schmelzpunkt: > 400 °C
Dichte: 11 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011045
CAS-Nummer: 21908-53-2
EINECS: 244-654-7
UN: 1641
ADR: 6.1 II
Merck Index: 13,05908

Bestellen

Spezifikation Testergebnisse

Assay (complexometric) ≥ 99.0 %
Substances insoluble in dilute hydrochloric acid ≤ 0.03 %
Chloride (Cl) ≤ 0.025 %
Sulfate (SO₄) ≤ 0.005 %
Total nitrogen (N) ≤ 0.005 %
Cd (Cadmium) ≤ 0.0001 %
Cu (Copper) ≤ 0.0005 %
Fe (Iron) ≤ 0.01 %
Pb (Lead) ≤ 0.0005 %
Zn (Zinc) ≤ 0.0005 %
Substances not reducing by formic acid (Residue on ignition, as sulfate) ≤ 0.025 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR