Quecksilber(II)sulfat, reinst, Supelco®
Lieferant: Merck
|
Gefahr
|
M-Clarity™ Qualitätsstufe = MQ200
1.04481.0100
1.04481.0250
1.04481.1000
1.04481.0100EA
200
EUR
1.04481.0100
1.04481.0250
1.04481.1000
Quecksilber(II)sulfat, reinst, Supelco®
Quecksilber(II)sulfat
Formel:
HgSO₄ Schmelzpunkt: 850 °C Dichte: 6,47 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011047 CAS-Nummer: 7783-35-9 EINECS: 231-992-5 UN: 1645 ADR: 6.1,II Merck Index: 13,05914 |
Spezifikation Testergebnisse
Identity | passes test |
Chloride (Cl) | ≤ 0.003 % |
Fe (Iron) | ≤ 0.005 % |
Mercury (I) (as Hg) | ≤ 0.1 % |
Substances not reducing by formic acid (residue on ignition) | ≤ 0.1 % |
Ignition Point |
|
Hazard Class | 6.1 |
Hazard Class Description | Toxic materials |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...