3-Methyl-2-benzothiazolinonehydrazone hydrochloride, GR zur Analyse, Supelco®

Lieferant: Merck
Gefahr

Synonyme: Sawicki's Reagent, 2-Hydrazono-3-methyl-2,3-dihydrobenzo[d]thiazole hydrochloride, MBTH Hydrochloride

M-Clarity™ Qualitätsstufe = MQ100

1.04527.0010
1.04527.0010EA 218 EUR
1.04527.0010
3-Methyl-2-benzothiazolinonehydrazone hydrochloride, GR zur Analyse, Supelco®
3-Methyl-2-benzothiazolinonehydrazone hydrochloride
Formel: C₈H₁₀ClN₃S
Molecular Weight: 215.70 g/mol
MDL: MFCD00005816
CAS-Nummer: 4338-98-1
UN: 2811
ADR: 6.1 III

Bestellen

Spezifikation Testergebnisse

Assay (argentometric) ≥98,5 %
Identity (IR-spectrum) Passes test
pH-value (0.4 %, water) 3 - 4
Melting point about 270°C
Sulfated ash ≤0,1 %
Loss on Drying (105°C) ≤ 1.0 %
Suitability for determination of aldehydes ≥0,62
Bulk density 748 kg/m3
Solubility Slightly soluble at 20 °C
Storage temperature +15...+25 °C
pH-value (0.4 %, water) 2,5 - 4,4
Loss on drying (105 °C) ≤1,0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR