Hydroxynaphtholblau Indikator für die Metalltitration, Supelco®

Lieferant: Merck

Synonyme: 1-(2-Hydroxy-4-sulpho-1-naphthylazo)-2-naphthol-3,6-disulphonic acid trisodium salt

M-Clarity™ Qualitätsstufe = MQ100

1.04593.0025
1.04593.0025EA 532 EUR
1.04593.0025
Hydroxynaphtholblau Indikator für die Metalltitration, Supelco®
Hydroxynaphthol blue
Formel: C₂₀H₁₁N₂Na₃O₁₁S₃
Molecular Weight: 620,47 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00036393
CAS-Nummer: 63451-35-4
EINECS: 264-197-7

Bestellen

Spezifikation Testergebnisse

Appearance of solution conforms
Loss on Drying (105 °C) ≤ 5 %
Suitability as indicator (for metal titration) conforms

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR