Eriochromblauschwarz R (Calcon®, Mordant Black 17), Supelco®
Lieferant: Merck
Synonyme:
1-(2-Hydroxy-1-naphthylazo)- 2-naphthol-4-sulfonsäure Natriumsalz, Natrium-3-hydroxy-4-(2-hydroxy-1-naphthylazo)-1-naphthalinsulfonat, Eriochromblauschwarz R, Palatinchromschwarz 6 BN, Calcon®
M-Clarity™ Qualitätsstufe = MQ200
1.04594.0050
1.04594.0050EA
64.7
EUR
1.04594.0050
Eriochromblauschwarz R (Calcon®, Mordant Black 17), Supelco®
Eriochromblauschwarz R (Calcon®, Mordant Black 17)
Formel:
C₂₀H₁₃N₂NaO₅S Molecular Weight: 416,38 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004069 CAS-Nummer: 2538-85-4 EINECS: 219-810-2 |
Spezifikation Testergebnisse
Solubility | 20 g/l (20 °C) |
Molar Mass | 416.39 g/mol |
Bulk Density | 530 kg/m3 |
pH | 9.4 (10 g/l, H2O, 25 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...