Hydraziniumsulfat, GR zur Analyse, Supelco®
Lieferant: Merck
|
Gefahr
|
M-Clarity™ Qualitätsstufe = MQ100
1.04603.0100
1.04603.0500
1.04603.0100EA
46.7
EUR
1.04603.0100
1.04603.0500
Hydraziniumsulfat, GR zur Analyse, Supelco®
Hydraziniumsulfat
Formel:
NH₂NH₂·H₂SO₄ Schmelzpunkt: 254 °C Dichte: 1,37 g/cm³ (20 °C) |
MDL:
MFCD00044873 CAS-Nummer: 10034-93-2 EINECS: 233-110-4 UN: 3288 ADR: 6.1,III Merck Index: 14,04772 |
|
Spezifikation Testergebnisse
Assay (iodometric) | ≥ 99.0% |
Identity | passes test |
Insoluble matter | ≤0.005 % |
Chloride (Cl) | ≤0.0005 % |
Heavy metals (as Pb) | ≤0.0005 % |
As (Arsenic) | ≤ 0.0001 % |
Fe (Iron) | ≤ 0.001 % |
Residue on ignition (as SO.) | ≤ 0.05 % |
Corresponds to ACS,Reag. Ph Eur |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...