Kaliumdihydrogenphosphat, kristallisiert, EMPROVE® ESSENTIAL Ph. Eur., BP, JPC, NF, E340, SAFC®
Lieferant: Merck
1.04871.1000
1.04871.5000
1.04871.9050
1.04871.9029
1.04871.1000EA
175
EUR
1.04871.1000
1.04871.5000
1.04871.9050
1.04871.9029
Kaliumdihydrogenphosphat, kristallisiert, EMPROVE® ESSENTIAL Ph. Eur., BP, JPC, NF, E340, SAFC®
Kaliumdihydrogenphosphat
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).
Standards: Corresponds to Ph Eur, BP, JPC, NF, E 340
Conforms to the purity criteria on food additives according to the current European Commission Regulation.
Formel:
KH₂PO₄ Siedepunkt: ∼100 °C (1013 hPa) Schmelzpunkt: 253 °C Dichte: 2,3 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011401 CAS-Nummer: 7778-77-0 EINECS: 231-913-4 REACH: 01-2119490224-41 Merck Index: 13,07744 |
Spezifikation Testergebnisse
Assay (alkalimetric, calculated on dried substance) | 98.0 - 100.5 % |
Assay (alkalimetric; dried substance) | ≥ 99.0 % |
Appearance of solution | passes test |
Identity | passes test |
Insoluble substances | ≤ 0.2 % |
pH-value (1 %; water) | 4.2 - 4.8 |
pH-value (2 %; water) | 4.2 - 4.6 |
pH-value (5 %; water) | 4.2 - 4.5 |
Carbonate | passes test |
Condensed phosphate | passes test |
Chloride (Cl) | ≤ 0.009 % |
Fluoride (F) | ≤ 0.0010 % |
Sulfate (SO₄) | ≤ 0.017 % |
Heavy metals (as Pb) | ≤ 0.0010 % |
Al (Aluminium) | ≤ 0.0005 % |
As (Arsenic) | ≤ 0.0001 % |
Cd (Cadmium) | ≤ 0.0001 % |
Fe (Iron) | ≤ 0.0010 % |
Hg (Mercury) | ≤ 0.0001 % |
Na (Sodium) | passes test |
Pb (Lead) | ≤ 0.0001 % |
Residual solvents (ICH Q3C) | excluded by manufacturing process |
Reducing substances | passes test |
Loss on drying (105 °C) | ≤ 0.5 % |
Loss on drying (130 °C) | ≤ 1.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...