Kaliumpermanganat, kristallin, EMPLURA®, Supelco®
Lieferant: Merck
|
Gefahr
|
Synonyme:
Permanganic acid potassium salt
M-Clarity™ Qualitätsstufe = MQ200
1.05080.1000
1.05080.5000
1.05080.9050
1.05080.1000EA
95.9
EUR
1.05080.1000
1.05080.5000
1.05080.9050
Kaliumpermanganat, kristallin, EMPLURA®, Supelco®
Potassium permanganate
Formel:
KMnO₄ Molecular Weight: 158.03 g/mol Schmelzpunkt: > 240 °C (Zers.) Dichte: 2.7 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011364 CAS-Nummer: 7722-64-7 EINECS: 231-760-3 UN: 1490 ADR: 5.1 II Merck Index: 12,07824 |
Spezifikation Testergebnisse
Assay (manganometric) | 99,0 - 100,5% |
Identity | passes test |
Appearance of solution | passes test |
Water insoluble matter | ≤ 0,2 % |
Chloride (Cl) | ≤ 0,01 % |
Sulphate (SO₄) | ≤ 0,03 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...