Kaliumpermanganat, kristallin, EMPLURA®, Supelco®

Lieferant: Merck
Gefahr

Synonyme: Permanganic acid potassium salt

M-Clarity™ Qualitätsstufe = MQ200

1.05080.1000 1.05080.5000 1.05080.9050
1.05080.1000EA 95.9 EUR
1.05080.1000 1.05080.5000 1.05080.9050
Kaliumpermanganat, kristallin, EMPLURA®, Supelco®
Potassium permanganate
Formel: KMnO₄
Molecular Weight: 158.03 g/mol
Schmelzpunkt: > 240 °C (Zers.)
Dichte: 2.7 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011364
CAS-Nummer: 7722-64-7
EINECS: 231-760-3
UN: 1490
ADR: 5.1 II
Merck Index: 12,07824

Bestellen

Spezifikation Testergebnisse

Assay (manganometric) 99,0 - 100,5%
Identity passes test
Appearance of solution passes test
Water insoluble matter ≤ 0,2 %
Chloride (Cl) ≤ 0,01 %
Sulphate (SO₄) ≤ 0,03 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR