Creatinin, Millipore®

Lieferant: Merck

Synonyme: 2-Amino-1-methyl-1,5-dihydro-4H-imidazol-4-one

MQ100

1.05206.0050 1.05206.0500
1.05206.0050EA 73.5 EUR
1.05206.0050 1.05206.0500
Creatinin, Millipore®
Creatinin
Formel: C₄H₇N₃O
Molecular Weight: 113.12 g/mol
MDL: MFCD00059730
CAS-Nummer: 60-27-5
EINECS: 200-466-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR