Kupfer-di-Ammonium -Lösung Cu(NH₄)₂-EDTA 0.1 mol/l, Titriplex®, Supelco®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
1.05217.0500
1.05217.0500EA
156
EUR
1.05217.0500
Kupfer-di-Ammonium -Lösung Cu(NH₄)₂-EDTA 0.1 mol/l, Titriplex®, Supelco®
Kupfer-di-Ammonium
Spezifikation Testergebnisse
Assay (Cu) | 0.098 - 0.102 mol/l |
pH-value | 9.7 - 10.3 |
Free EDTA | conforms |
Free copper | conforms |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...