Dowex® 50WX8 (H) (stark saurer Kationenaustauscher), Supelco®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

1.05221.0250
1.05221.0250EA 373 EUR
1.05221.0250
Dowex® 50WX8 (H) (stark saurer Kationenaustauscher), Supelco®
Dowex® 50WX8 (H)
MDL: MFCD00132726
CAS-Nummer: 11119-67-8

Bestellen

Spezifikation Testergebnisse

Description light yellow to brown spherules
Particle size (0,09-0,25 mm) ≥ 70.0 %
Total capacity ≥ 1.7 mol/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR