Ölrot O, Certistain® für die Mikroskopie, Sigma-Aldrich®

Lieferant: Merck

Synonyme: CI 26125

M-Clarity™ Qualitätsstufe = MQ300

1.05230.9999
1.05230.9999EA 0 EUR
1.05230.9999
Ölrot O, Certistain® für die Mikroskopie, Sigma-Aldrich®
Ölrot O

The CERTISTAIN® dry dye Oil red O (C.I. 26125) is used for the preparation of a staining solution that can be used for the detection of lipids in histological cryo sections.
As CERTISTAIN® dye it is chemically quality-certified according to highest standards and strict specifications. As an IVD product and CE registered, it can be used hassle-free in diagnostics and laboratory accreditations.

Formel: C₂₆H₂₄N₄O
Molecular Weight: 408,51 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00003898
CAS-Nummer: 1320-06-5
EINECS: 215-295-3
IVDLogo

Bestellen

Spezifikation Testergebnisse

Dye content (spectrophotometrically) ≥ 80 %
Identity (UV/VIS-Spectrum) passes test
Absorption maximum λ max (chloroform) 528 - 534 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.01 g/l; chloroform) 725 - 910
TLC-Test passes test
Loss on drying (110 °C) ≤ 8 %
Suitability for microscopy passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR