L(+)-Isoleucin, EMPROVE® ESSENTIAL Ph. Eur., USP, SAFC®

Lieferant: Merck

1.05357.9010 1.05357.1000
1.05357.9010EA 0 EUR
1.05357.9010 1.05357.1000
L(+)-Isoleucin, EMPROVE® ESSENTIAL Ph. Eur., USP, SAFC®
L(+)-Isoleucin
Formel: H₃CCH₂CH(CH₃)CH(NH₂)COOH
Schmelzpunkt: 279…280 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00064222
CAS-Nummer: 73-32-5
EINECS: 200-798-2
Merck Index: 14,05179

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR