L(+)-Isoleucin, Millipore®

Lieferant: Merck

Synonyme: (2S,3S)-(+)-2-Amino-3-methylpentansäure, (2S,3S)-(+)-α-Amino-β-methylvaleriansäure, (2S,3S)-(+)-Isoleucin

MQ100

1.05362.0500
1.05362.0500EA 523 EUR
1.05362.0500
L(+)-Isoleucin, Millipore®
L(+)-Isoleucin
Formel: H₃CCH₂CH(CH₃)CH(NH₂)COOH
Molecular Weight: 131.17 g/mol
Schmelzpunkt: 279…280 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00064222
CAS-Nummer: 73-32-5
EINECS: 200-798-2
Merck Index: 14,05179

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR