Leishman's Eosin-Methylenblau modifizierte Lösung Färbemittel für Histologie, Sigma-Aldrich®

Lieferant: Merck
Gefahr

Synonyme: Leishmans Eosin-Methylenblau, Leishman's Farbstoff

1.05387.0500 1.05387.0000 1.05387.1022
1.05387.0500EA 103 EUR
1.05387.0500 1.05387.0000 1.05387.1022
Leishman's Eosin-Methylenblau modifizierte Lösung Färbemittel für Histologie, Sigma-Aldrich®
Leishman's Eosin-Methylenblau

For staining of blood and bone marrow smears and clinical-cytological specimens


The staining method according to Leishman is one of the standard techniques in hematological diagnostic procedures. Like Wright's eosin methylene blue solution and May-Grünwald's eosin methylene blue solution, it is used for staining of blood and bone marrow smears and clinical cytological specimens. The typical resulting color of cell nuclei is red to purple, which depends on the buffer tablets (Buffer tablets pH 7.2 or Buffer tablets pH 6.8). 500 mL of the IVD & CE-registered Leishman's eosin methylene blue solution are sufficient for staining up to 1600 slides in Hellendahl cells. It is registered as IVD and CE product.


Preparative capacity: 1000 ml


3200 slides automate/staining cell (diluted)
500 slides staining rack (undiluted)

Siedepunkt: 65 °C (1013 hPa)
Dichte: 0,79 g/cm3 (20 °C)
Flammpunkt: 12 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00131498
CAS-Nummer: 12627-53-1
UN: 1230
ADR: 3 (6.1),II
IVDLogo

Bestellen

Spezifikation Testergebnisse

Suitability for microscopy (Blood smear) passes test
Erythrocytes reddish
Nuclei violet
Eosinophilic granules brick-red
Neutrophilic granules light violet
Lymphocyte cytoplasm Blue


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR