Ammoniaklösung 25%, EMPROVE® ESSENTIAL Ph. Eur., BP, reinst suitable for use as excipient, SAFC®

Lieferant: Merck
Gefahr

Synonyme: Ammoniumhydroxid, Ammoniak

1.05422.1000 1.05422.2500 1.05422.9025 1.05422.9180
1.05422.1000EA 64.3 EUR
1.05422.1000 1.05422.2500 1.05422.9025 1.05422.9180
Ammoniaklösung 25%, EMPROVE® ESSENTIAL Ph. Eur., BP, reinst suitable for use as excipient, SAFC®

Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).


Standards: Conforms to Ph Eur, BP

Formel: NH₃
Siedepunkt: 37,7 °C (1013 hPa)
Schmelzpunkt: -57,5 °C
Dichte: 0,903 g/cm3 (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00066650
CAS-Nummer: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8,III

Bestellen

Spezifikation Testergebnisse

Assay (acidimetric, NH₃) 25.0 - 30.0 %
Identity passes test
Appearance of solution passes test
Density (20/20°C) 0.892 - 0.910
Carbonate (as CO₂) ≤ 0.0060 %
Chloride (Cl) ≤ 0.00010 %
Sulfate (SO₄) ≤ 0.0005 %
Pyridine and related substances ≤ 0.0002 %
Cu (Copper) ≤ 0.00005 %
Fe (Iron) ≤ 0.000025 %
Oxidizable substances passes test
Substances reducing potassium permanganate (as O) ≤ 0.00075 %
Residual solvents (ICH Q3C) ...excluded by production process
Evaporation residue ≤ 0.0018 %
Residue on ignition (as SO₄) ≤ 0.002 %
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*). ≤ 0.002 %
Conforms to Ph Eur, BP


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR