Kaliumhydroxid 0.1 mol/l (0.1 N) isopropanol, Titripur® Volumetric acc. to DIN 51558 part 1, Supelco®

Lieferant: Merck
Gefahr

Synonyme: Kalihydrat, Ätzkali

MQ100

1.05544.1000 1.05544.1003
1.05544.1000EA 49.11 EUR
1.05544.1000 1.05544.1003
Kaliumhydroxid 0.1 mol/l (0.1 N) isopropanol, Titripur® Volumetric acc. to DIN 51558 part 1, Supelco®
Kaliumhydroxid

Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.

Formel: KOH
Molecular Weight: 56.11 g/mol
Siedepunkt: 1320
Schmelzpunkt: 360
Lagertemperatur: Raumtemperatur
MDL: MFCD00003553
CAS-Nummer: 1310-58-3
EINECS: 215-181-3
UN: 2924
ADR: 3 (8) II
Merck Index: 12,07806

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR