tri-Lithiumcitrat Tetrahydrat, EMPROVE® ESSENTIAL BP, Ph. Eur. suitable for use as excipient, SAFC®

Lieferant: Merck
Achtung

Synonyme: Citronensäure Trilithiumsalz Tetrahydrat, Trilithium-2-hydroxypropan-1,2,3-tricarboxylat Tetrahydrat, Lithiumcitrat tribasisch Tetrahydrat

1.05683.9024
1.05683.9024EA 0 EUR
1.05683.9024
tri-Lithiumcitrat Tetrahydrat, EMPROVE® ESSENTIAL BP, Ph. Eur. suitable for use as excipient, SAFC®
tri-Lithiumcitrat Tetrahydrat
Formel: C₆H₅Li₃O₇·4H₂O
Molecular Weight: 281.99 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00150030
CAS-Nummer: 6080-58-6
EINECS: 213-045-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR