L(+)-Lysin Monohydrochlorid, Millipore®

Lieferant: Merck

Synonyme: (S)-(+)-Lysin Monohydrochlorid, H-Lys-OH.HCl, (S)-(+)-2,6-Diaminohexansäure Hydrochlorid, (S)-(+)-2,6-Diaminocapronsäure Hydrochlorid, L(+)-Lysin Hydrochlorid, (S)-(+)-2,6-Diaminohexansäure Monohydrochlorid, (S)-(+)-2,6-Diaminocapronsäure Monohydrochlorid, (S)-(+)-Lysin Hydrochlorid

M-Clarity™ Qualitätsstufe = MQ100

1.05700.1000 1.05700.9010
1.05700.1000EA 214 EUR
1.05700.1000 1.05700.9010
L(+)-Lysin Monohydrochlorid, Millipore®
L(+)-Lysin Monohydrochlorid
Formel: H₂N(CH₂)₄CH(NH₂)CO₂H·HCl
Schmelzpunkt: 263…264 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00064564
CAS-Nummer: 657-27-2
EINECS: 211-519-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR