Molekularsieb A4 (0,4 nm, 4 Å), Stäbe ~ 1.6 mm (1/16"), Supelco®
Lieferant: Merck
Synonyme:
Molecular sieve 0.4 nm, Molecular sieves pack 4A, Linde 4A, Sodium aluminium silicate
M-Clarity™ Qualitätsstufe = MQ200
1.05743.1000
1.05743.1000EA
233
EUR
1.05743.1000
Molekularsieb A4 (0,4 nm, 4 Å), Stäbe ~ 1.6 mm (1/16"), Supelco®
Molekularsieb A4 (0,4 nm, 4 Å)
Dichte:
1,1…2.2 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD03457537 CAS-Nummer: 70955-01-0 EINECS: 215-283-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...