Maltose Monohydrat ≥95,0% (durch HPLC), EMPROVE® ESSENTIAL, SAFC®

Lieferant: Merck

Synonyme: Malzzucker

1.05911.9029
1.05911.9029EA 4390 EUR
1.05911.9029
Maltose Monohydrat ≥95,0% (durch HPLC), EMPROVE® ESSENTIAL, SAFC®
D(+)-Maltose Monohydrat

Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).

Formel: C₁₂H₂₂O₁₁·1H₂O
Molecular Weight: 360,32 g/mol
Schmelzpunkt: 160…165 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00149343
CAS-Nummer: 6363-53-7
EINECS: 200-716-5
Merck Index: 13,05736
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f861d7adf53134a0c485b5942d3be34c4d0b571&vid=16e5c4425a7a8e158a4200470a45483c7ff5781e&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Purity (HPLC) ≥ 95.0 %
Identity (IR) passes test
Appearance of solution (clearness) passes test
Appearance of solution (colour) passes test
pH-value (5 %; water; 20°C) 4.5 - 6.0
Spec. rotation [α²0/D (10 %; water; calc. on anhydrous substance) 135.0 - 139.0 °
Heavy metals (as Pb) ≤ 0.002 %
Residual solvents (ICH Q3C) excluded by manufacturing process
Sulfated ash ≤ 0.1 %
Water 4.0 - 7.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR