(-)-Menthol, EMPROVE® ESSENTIAL BP, Ph. Eur., JP, USP suitable for use as excipient, SAFC®

Lieferant: Merck
Achtung

Synonyme: L(-)-Menthylalkoholol, (1R)-(-)-Menthol, Levomenthol, (1R,2S,5R)-(-)-Menthol, (1α,2β,4β)-(-)-2-Hydroxy-1-isopropyl-4-methylcyclohexan, (1S,2R,4R)-(-)-2-Hydroxy-1-isopropyl-4-methylcyclohexan

1.05995.1000 1.05995.9025
1.05995.1000EA 545 EUR
1.05995.1000 1.05995.9025
(-)-Menthol, EMPROVE® ESSENTIAL BP, Ph. Eur., JP, USP suitable for use as excipient, SAFC®
L(-)-Menthol
Formel: C₁₀H₂₀O
Siedepunkt: 212 °C (1013 hPa)
Schmelzpunkt: 41…44 °C
Dichte: 0,89 g/cm³ (20 °C)
Flammpunkt: 96 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00062979
CAS-Nummer: 2216-51-5
EINECS: 218-690-9
Merck Index: 13,05861

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR