Methanol ≥99,9% (durch GC), LiChrosolv® Reag. Ph. Eur., Gradienten-Qualität für die Flüssigchromatographie, Supelco®

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ200

1.06007.1000 1.06007.2500 1.06007.4000 1.06007.9010 1.06007.9030 1.06007.9185 1.06007.5000 1.06007.9912
1.06007.1000EA 56.7 EUR
1.06007.1000 1.06007.2500 1.06007.4000 1.06007.9010 1.06007.9030 1.06007.9185 1.06007.5000 1.06007.9912
Methanol ≥99,9% (durch GC), LiChrosolv® Reag. Ph. Eur., Gradienten-Qualität für die Flüssigchromatographie, Supelco®
Methanol

With their high degree of UV transmittance, low particle count, low acidity and alkalinity and low evaporation residue level, LiChrosolv® solvents are ideal for reproducible separations. Since separations are normally carried out under gradient conditions in analytical HPLC, we offer solvents in "gradient grade" as well as "isocratic grade". This enables to minimize the gradient effect of the solvent involved.


  • Filtered by 0,2 µm filter
  • Suitable for UPLC / UHPLC / Ultra HPLC - instruments
  • Conforms to Methanol R1 and R2 according to Reag. Ph. Eur.
  • Conforms to the requirements of ACS for liquid chromatography suitability


With their high degree of UV transmittance, low particle count, low acidity and alkalinity and low evaporation residue level, LiChrosolv® solvents are ideal for reproducible separations. Since separations are normally carried out under gradient conditions in analytical HPLC, we offer solvents in a 'gradient grade' as well as 'isocratic grade'. This minimises the gradient effect of the solvent involved.

Formel: CH₃OH
Siedepunkt: 64,6 °C (1013 hPa)
Schmelzpunkt: –98 °C
Dichte: 0,7918 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004595
CAS-Nummer: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3 (6.1),II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Purity (GC) ≥99.9%
Identity (IR) Conforms
Evaporation residue ≤2.0 mg/l
Water ≤0.02%
Colour ≤10 Hazen
Acidity ≤0.0002 meq/g
Alkalinity ≤0.0002 meq/g
Gradient grade (at 235 nm) ≤2.0 mAU
Gradient grade (at 254 nm) ≤1.0 mAU
Fluorescence (as quinine at 254 nm) ≤1.0 ppb
Fluorescence (as quinine at 365 nm) ≤0.5 ppb
Transmission (at 210 nm) ≥20%
Transmission (at 220 nm) ≥60%
Transmission (at 230 nm) ≥75%
Transmission (at 235 nm) ≥83%
Transmission (at 250 nm) ≥95%
Transmission (from 260 nm) ≥98%
Absorbance (at 225 nm) ≤0.170


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR