Methylrot, Supelco®
Lieferant: Merck
Synonyme:
2-(4-(Dimethylamino)phenylazo)benzoesäure
M-Clarity™ Qualitätsstufe = MQ200
1.06076.0025
1.06076.0100
1.06076.0025EA
61.4
EUR
1.06076.0025
1.06076.0100
Methylrot, Supelco®
Methylrot
Formel:
C₁₅H₁₅N₃O₂ Molecular Weight: 269.3 g/mol Schmelzpunkt: 178…182 °C Dichte: 1.31 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002425 CAS-Nummer: 493-52-7 EINECS: 207-776-1 Merck Index: 12,06199 |
Spezifikation Testergebnisse
Solubility | (20 °C) slightly soluble |
Melting Point | 178 - 182 °C |
Molar Mass | 269.31 g/mol |
Bulk Density | 300 - 500 kg/m3 |
Hazard Class | 9 |
Hazard Class Description | Miscellaneous Hazardous Material |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...