CAPSO (3-(Cyclohexylamino)-2-hydroxypropansulfonsäure)
Lieferant: Merck
Synonyme:
3-(Cyclohexylamino)-2-hydroxy-1-propanesulphonic acid
1.06186.0100
1.06186.0100EA
0
EUR
1.06186.0100
CAPSO (3-(Cyclohexylamino)-2-hydroxypropansulfonsäure)
CAPSO (3-(Cyclohexylamino)-2-hydroxypropansulfonsäure)
Formel:
C₉H₁₉NO₄S Molecular Weight: 237.32 g/mol Schmelzpunkt: 270…274 °C |
MDL:
MFCD00041778 CAS-Nummer: 73463-39-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...