di-Natriumhydrogenphosphat Dodecahydrat 98,5-102,5% (durch Acidimetrie), Kristalle, EMSURE® Ph. Eur., JP, USP analytical reagent, Supelco®

Lieferant: Merck

Synonyme: Natriumphosphat dibasisch Dodecahydrat, sek. Natriumphosphat Dodecahydrat

M-Clarity™ Qualitätsstufe = MQ300

1.06579.0500 1.06579.1000 1.06579.5000 1.06579.9025
1.06579.0500EA 49.3 EUR
1.06579.0500 1.06579.1000 1.06579.5000 1.06579.9025
di-Natriumhydrogenphosphat Dodecahydrat 98,5-102,5% (durch Acidimetrie), Kristalle, EMSURE® Ph. Eur., JP, USP analytical reagent, Supelco®
di-Natriumhydrogenphosphat Dodecahydrat
Formel: Na₂HPO₄·12H₂O
Molecular Weight: 358.14 g/mol
Schmelzpunkt: 35
Dichte: 1.52 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149181
CAS-Nummer: 10039-32-4
EINECS: 231-448-7
Merck Index: 13,08733

Bestellen

Spezifikation Testergebnisse

Assay (alkalimetric) 99.0 - 102.0 %
Identity passes test
Appearance of solution passes test
Sodium Dihydrogene phospate ≤ 2.5 %
pH-value (5 %; water) 9.0 - 9.3
Chloride (Cl) ≤ 0.0005 %
Sulfate (SO₄) ≤ 0.005 %
Total nitrogen (N) ≤ 0.001 %
Heavy metals (as Pb) ≤ 0.0005 %
As (Arsenic) ≤ 0.00005 %
Cu (Copper) ≤ 0.0002 %
Fe (Iron) ≤ 0.0005 %
K (Potassium) ≤ 0.005 %
Pb (Lead) ≤ 0.0005 %
Reducing substances passes test
Loss on drying (130 °C) 57.0 - 61.0 %
Corresponds to ISO,Reag. Ph Eur passes test
Water (according to Karl Fischer) 57.0 - 61.0 %
Corresponds to Ph Eur, BP, USP, JP


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR