di-Natriumhydrogenphosphat, wasserfrei ≥99,0%, EMSURE® ACS, Reag. Ph. Eur. zur Analyse, Supelco®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ300
1.06586.0500
1.06586.2500
1.06586.9050
1.06586.1000
1.06586.9012
1.06586.0500EA
75.6
EUR
1.06586.0500
1.06586.2500
1.06586.9050
1.06586.1000
1.06586.9012
di-Natriumhydrogenphosphat, wasserfrei ≥99,0%, EMSURE® ACS, Reag. Ph. Eur. zur Analyse, Supelco®
di-Natriumhydrogenphosphat
Standards: Corresponds to ACS, Reag. Ph Eur
Formel:
Na₂HPO₄ Schmelzpunkt: 250 °C Dichte: 1,53 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003496 CAS-Nummer: 7558-79-4 EINECS: 231-448-7 REACH: 01-2119489797-11 Merck Index: 12,08805 |
Spezifikation Testergebnisse
Assay (alkalimetric) | ≥99.0% |
Insoluble matter | ≤0.01% |
pH-value (5%; water, 25 °C) | 8.7 - 9.3 |
Chloride (Cl) | ≤0.002% |
Sulfate (SO₄) | ≤0.005% |
Heavy metals (as Pb) | ≤0.001% |
Cu (Copper) | ≤0.0003% |
Fe (Iron) | ≤0.001% |
K (Potassium) | ≤0.01% |
Pb (Lead) | ≤0.001% |
Loss on Drying (105 °C) | ≤0.2% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...