Natriumammoniumhydrogenphosphat Tetrahydrat, EMSURE® zur Analyse, Supelco®
Lieferant: Merck
Synonyme:
XSodium ammonium hydrogen phosphate tetrahydrate, Sodium ammonium phosphate, Ammonium sodium phosphate dibasic tetrahydrate, Sodium ammonium phosphate tetrahydrate, Ammonium sodium phosphate tetrahydrate
M-Clarity™ Qualitätsstufe = MQ300
1.06682.1000
1.06682.1000EA
325
EUR
1.06682.1000
Natriumammoniumhydrogenphosphat Tetrahydrat, EMSURE® zur Analyse, Supelco®
Sodium ammonium hydrogen phosphate tetrahydrate
Formel:
NaNH₄HPO₄·4 H₂O Molecular Weight: 209,07 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00066970 CAS-Nummer: 7783-13-3 EINECS: 235-860-8 |
Spezifikation Testergebnisse
Solubility | 167 g/l (20 °C) |
Melting Point | 79 °C |
Molar Mass | 209.07 g/mol |
Density | 0.85 g/cm3 (20 °C) |
pH | 8.3 (50 g/l, H2O, 20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...