Papanicolaou's Lösung 2a Orange G (OG 6), Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
Cytoplasm staining of mature and keratinized cells in cytology
The effect of the orange staining solution is considerable in smears with keratinized cells under acidic pH conditions, when the obvious orangeophilic cytoplasmic stain is recognizable and may point to the presence of hyperkeratosis, HPV infection or carcinoma cells. Papanicolaou's solution 2a, Orange G solution gives a gentle yellow-orange staining result with mature and keratinized squamous cells. Papanicolaou's solution 2b, Orange II solution gives a intense reddish staining result with mature and keratinized squamous cells. A 500 ml bottle is sufficient for 1500 to 2000 stainings.
Dichte:
0,83 g/cm³ (20 °C) Flammpunkt: 14 °C |
UN:
1170 ADR: 3,II |
![]() |
Spezifikation Testergebnisse
Suitability for microscopy (vaginal smear) | Passes test |
Nuclei | Blue to dark violet |
Cyanophilic cytoplasm | Blue-green |
Eosinophilic cytoplasm | Pink |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...