Silica gel, Geduran® Si 60, for high performance flash chromatography
Lieferant: Merck
This silica gel is suitable for high performance flash chromatography.
Boiling point: 2230 °C
Melting Point: 1710 °C
pH value: 7
Bulk density: 200 – 1430 kg/m³
Verpackung: 25 kg/pack in plastic drum.