Paraffin, Erstarrungspunkt: 56…58 °C, pastille Histology, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Wax Paraffin, Paraffin wax
1.07164.2504
1.07164.2504EA
250
EUR
1.07164.2504
Paraffin, Erstarrungspunkt: 56…58 °C, pastille Histology, Sigma-Aldrich®
Paraffin
Paraffin is a traditional embedding media for histology
In order to achieve uniformous ribbon-sectioning quality, defined hardness and homogeneity of the processed material is necessary. This can be guarranteed through high quality raw materials and defined melting points. The melting point of paraffin is 56 to 58 °C. It is registered as IVD and CE certified product and can be used for clinical diagnostics.
Dichte:
0.9 g/cm³ (20 °C) Flammpunkt: >100 °C Lagertemperatur: Raumtemperatur |
CAS-Nummer:
64742-51-4 EINECS: 232-315-6 |
![]() |
Spezifikation Testergebnisse
Effect of heating (at 60 °C/100 g) | passes test |
Solubility in xylene (at 20 °C/0.1 g in 5 ml) | passes test |
Cutting results (4 μm slices) | passes test |
Solubility of slices in xylene | ≤ 50 s |
Solidification point (rotating thermometer method) | 56 - 58°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...