Peracetic acid ∼38 - 40%, Supelco®

Lieferant: Merck
Gefahr

Synonyme: Peroxyacetic acid

M-Clarity™ Qualitätsstufe = MQ200

1.07222.1000
1.07222.1000EA 189 EUR
1.07222.1000
Peracetic acid ∼38 - 40%, Supelco®
Peracetic acid
Formel: C₂H₄O₃
Molecular Weight: 76,051 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00002128
CAS-Nummer: 79-21-0
UN: 3105
ADR: 5.2 (8),

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR