n-Pentan ≥99,9% (durch GC), Suprasolv® hypergrade für die organische Spurenanalyse, Supelco®

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ100

1.07288.1000 1.07288.2500
1.07288.1000EA 143 EUR
1.07288.1000 1.07288.2500
n-Pentan ≥99,9% (durch GC), Suprasolv® hypergrade für die organische Spurenanalyse, Supelco®
n-Pentan

SupraSolv® ECD and FID is specially developed for gas chromatography in combination with ECD (Electron Capture Detetor) and FID (Flame Ionization detector). SupraSolv® solvents offer the largest specified retention time range, a clear baseline and a minimal signal-to-noise ratio leading to reliable and reproducible analysis results.

Formel: H₃C(CH₂)₃CH₃
Siedepunkt: 35…36 °C (1013 hPa)
Schmelzpunkt: –130 °C
Dichte: 0,63 g/cm³ (20 °C)
Flammpunkt: –48 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00009498
CAS-Nummer: 109-66-0
EINECS: 203-692-4
UN: 1265
ADR: 3,II
REACH: 01-2119459286-30
Merck Index: 13,07193
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ba2c9c81a4b1f44769627a00ba49b6cc8e63132&vid=f6831eb15ab1602d8c752468dc3ee31cc4ee61c0&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Purity (GC) ≥ 99.9%
Identity (IR) Conforms
Evaporation Residue ≤3.0 mg/L
Water ≤0.01%
Color ≤10 Hazen
GC/ECD (Retention Range 1,2,4-tri- Chlorobenzene to Decachlorobiphenyle, Individual Signals (Lindane Standard)) ≤2 pg/ml
GC/ECD (Retention Range Dichloromethane to 1,2,4-Trichlorobenzene Individual Signals (Tetrachloromethane)) ≤1 ng/ml
GC/FID (Retention Range n-Undecane to n-Tetracontane, Individual Signals (n-Tetradecane Standard)) ≤2 ng/ml
GC/MSD (Retention Range n-Undecane to n-Tetracontane, Scaning area 30 - 600 amu, Individual Signals (n- Tetradecane Standard)) ≤2 ng/ml
Suitable for Residue Analysis


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR