Polyvinylpyrrolidon, Divergan® RS, Supelco®

Lieferant: Merck

Synonyme: Polyvidon, Poly[1-(2-oxo-1-pyrrolidinyl)ethylen], PVP

M-Clarity™ Qualitätsstufe = MQ100

1.07302.0100
1.07302.0100EA 112 EUR
1.07302.0100
Polyvinylpyrrolidon, Divergan® RS, Supelco®
Polyvinylpyrrolidon

Polyvinylpolypyrrolidone is used as excipient. It has a wide range of applications. In connection with the Reflectoquant®- systems it is applicable in the beverage industry.


In the case of some colored samples, it may be necessary to decolorize the solution with polyvinylpolypyrrolidone prior to the measurement, at the same time leaving the constituent of the product under analysis unchanged. The volume, type, and application time of the Divergan® product have a major effect on the decolorization process, meaning that the dose of Divergan® must be carefully adjusted to the analyte under investigation.

Formel: (C₆H₉NO)n
Schmelzpunkt: 220 °C (Zers.)
Dichte: 1,2 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD01076626
CAS-Nummer: 9003-39-8

Bestellen

Spezifikation Testergebnisse

Identity (IR spectrum) conforms
Molar Mass 40000 g/mol
Bulk Density 330 kg/m3
Density 1.2 g/cm3 (20 °C)
Ignition Point 440 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR