Blei(II)fluorid, anhydrous 99,9, powder, Optipur®

Lieferant: Merck
Gefahr

Synonyme: Lead difluoride

1.07386.9010
1.07386.9010EA 0 EUR
1.07386.9010
Blei(II)fluorid, anhydrous 99,9, powder, Optipur®
Blei(II)fluorid

Optipur® materials are inorganic compounds, primarily metal halides, and available as anhydrous powders, beads or pieces for immediate usability and easy handling. They are of the highest purity, and specifically suited for the production of single crystals, used in demanding optical applications or scintillators, for example medical imaging and radiation detection.


  • High purity
  • Anhydrous powders or beads
  • Mainly metal halides
  • Immediate usability and easy handling
  • Special packaging for hygroscopic materials


The purity of each production batch is strictly controlled with careful attention paid to impurities that have an influence on transmission, light scattering, or scintillation efficiency. Hygroscopic materials are typically vacuum-dried at high temperature and are packaged for dry storage.

Formel: F₂Pb
Molecular Weight: 245.2 g/mol
Siedepunkt: 1293 °C (1013 hPa)
Schmelzpunkt: 824 °C
MDL: MFCD00011162
CAS-Nummer: 7783-46-2
UN: 2291
ADR: 6.1 III
Merck Index: 12,05425

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR