Pronase E (from Streptomyces griseus)
Lieferant: Merck
1.07433.0001
1.07433.0005
1.07433.0001EA
162
EUR
1.07433.0001
1.07433.0005
Pronase E (from Streptomyces griseus)
Enzyme
Pronase E is a mixture of proteolytic enzymes that is obtained from Streptomyces griseus. It can be used in the in diagnostics in histopathology as it hydrolyses proteins into individual amino acids.