1,2-Propandiol, EMPROVE® EXPERT BP, Ph. Eur., USP suitable for use as excipient, SAFC®
Lieferant: Merck
Synonyme:
Propylenglycol, (±)-1,2-Propylenglycol, (±)-1,2-Dihydroxypropan
1.07478.2500
1.07478.6025
1.07478.2500EA
362
EUR
1.07478.2500
1.07478.6025
1,2-Propandiol, EMPROVE® EXPERT BP, Ph. Eur., USP suitable for use as excipient, SAFC®
Propylenglycol
Formel:
CH₃CH(OH)CH₂OH Molecular Weight: 76.1 g/mol Siedepunkt: 187.6 Schmelzpunkt: –59 °C Dichte: 1.04 g/cm³ (20 °C) Flammpunkt: 99 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064272 CAS-Nummer: 57-55-6 EINECS: 200-338-0 Merck Index: 13,07947 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...