D(+)-Saccharose, Millipore®
Lieferant: Merck
Synonyme:
Haushaltszucker, D(+)-Sucrose, Rübenzucker, Rohrzucker, 2-β-D-Fructofuranosyl-1-α-D-glucopyranosid, Sucrose, Kristallzucker
MQ100
1.07651.9025
1.07651.9025EA
0
EUR
1.07651.9025
D(+)-Saccharose, Millipore®
D(+)-Saccharose
Formel:
C₁₂H₂₂O₁₁ Molecular Weight: 342.3 g/mol Schmelzpunkt: 169…170 °C Dichte: 1.29 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006626 CAS-Nummer: 57-50-1 EINECS: 200-334-9 Merck Index: 13,08966 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...