Lactose Monohydrat, powder, EMPROVE® EXPERT BP, Ph. Eur., JP, NF excipient_suitable, SAFC®

Lieferant: Merck

Synonyme: 4-(β-D-Galactopyranosyl)-D-glucopyranose Monohydrat, 4-(β-D-Galactopyranosyl)-D-glucopyranose 1-Hydrat, 4-(β-D-Galactopyranosyl)-D-glucopyranose-1-Hydrat, 4-(β-D-Galactopyranosyl)-D-glucopyranose-Monohydrat

1.07656.9029 1.07656.9525
1.07656.9029EA 0 EUR
1.07656.9029 1.07656.9525
Lactose Monohydrat, powder, EMPROVE® EXPERT BP, Ph. Eur., JP, NF excipient_suitable, SAFC®
Lactose Monohydrat
Formel: C₁₂H₂₂O₁₁·1H₂O
Molecular Weight: 360.32 g/mol
Schmelzpunkt: 223
Lagertemperatur: Raumtemperatur
CAS-Nummer: 10039-26-6
EINECS: 200-559-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR