Adhesion promoter, Silane A 174
Lieferant: Merck
1.07673.0050
1.07673.0050EA 121 EUR
1.07673.0050
Adhesion promoter, Silane A 174
Klebstoffe
Silane A 174 (also known as Polyfix® 100, methacryloxypropyl trimethoxysilane) is an adhesion promoter. Has a molar mass 248,35 g/mol.