Tetrahydrofuran, EMPLURA®, reinst, Supelco®

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ200

1.08114.1000 1.08114.2500 1.08114.6025 1.08114.6190 1.08114.9190
1.08114.1000EA 68.1 EUR
1.08114.1000 1.08114.2500 1.08114.6025 1.08114.6190 1.08114.9190
Tetrahydrofuran, EMPLURA®, reinst, Supelco®
Tetrahydrofuran

EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA® we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA® solvents provide adequate specifications with the most important parameters.

Formel: C₄H₈O
Siedepunkt: 66 °C (1013 hPa)
Schmelzpunkt: –108,5 °C
Dichte: 0,888 g/cm³ (20 °C)
Flammpunkt: –21,5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005356
CAS-Nummer: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3,II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR