Tetramethylammoniumhydroxid 10% aqueous solution Reag. Ph. Eur. polarography, examination of steroids, Supelco®
Lieferant: Merck
|
Gefahr
|
MQ100
1.08123.0050
1.08123.0250
1.08123.0050EA
85.8
EUR
1.08123.0050
1.08123.0250
Tetramethylammoniumhydroxid 10% aqueous solution Reag. Ph. Eur. polarography, examination of steroids, Supelco®
Tetramethylammonium hydroxide
for polarography and for examination of steroids
Formel:
C₄H₁₃NO Molecular Weight: 91.15 g/mol Dichte: 1.02 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008280 CAS-Nummer: 75-59-2 EINECS: 200-882-9 UN: 1835 ADR: 8 II Merck Index: 14,09224 |
Spezifikation Testergebnisse
Assay (acidimetric) | ≥ 10.0 % |
Chloride (Cl) | ≤ 0.004 % |
Ammonia and other amines | ≤ 0.2 % |
Ag (Silver) | ≤ 0.001 % |
Fe (Iron) | ≤ 0.001 % |
Pb (Lead) | ≤ 0.001 % |
Evaporation residue | passes test |
Ash | ≤ 0.05 % |
Suitability for polarography | passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...