Thymolphthalein ACS, Reag. Ph. Eur. Indikator
Lieferant: Merck
Synonyme:
2-(1-Hydroxy-1,1-di(4-hydroxy-2-methyl-5-(methylethyl)phenyl)methyl)benzoesäurelacton, 3,3-Bis(4-hydroxy-5-isopropyl-o-tolyl)phthalid
M-Clarity™ Qualitätsstufe = MQ200
1.08175.0005
1.08175.0025
1.08175.0100
1.08175.0005EA
72.4
EUR
1.08175.0005
1.08175.0025
1.08175.0100
Thymolphthalein ACS, Reag. Ph. Eur. Indikator
Thymolphthalein
Formel:
C₂₈H₃₀O₄ Schmelzpunkt: 253 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005909 CAS-Nummer: 125-20-2 EINECS: 204-729-7 Merck Index: 12,09542 |
Spezifikation Testergebnisse
Appearance | White to almost white powder, eventually slightly yellowish. |
Identity (UV/VIS-Spectrum) | passes test |
Absorption maximum λmax. (buffer pH 10.5) | 592 - 596 nm |
Spec. Absorptivity A 1%/1cm (λmax; 0.01 g/l; buffer pH 10.5; calculated on anhydrous substance) | 800 - 900 |
Transition range (according to ACS) | passes test |
Transition range | pH 9.0 - pH 10.5 colourless - blue |
Loss on drying (110 °C) | ≤ 1 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...