Cyclopentylmethylether (CPME), EMPLURA®

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ200

1.08293.1000 1.08293.2500 1.08293.4000
1.08293.1000EA 123 EUR
1.08293.1000 1.08293.2500 1.08293.4000
Cyclopentylmethylether (CPME), EMPLURA®
Cyclopentylmethylether (CPME)

Cyclopentyl methyl ether is a safer substitute for tetrahydrofuran, tert-butyl methyl ether, 1,4 dioxane and other ether solvents. It is produced by a 100% atomic catalytic reaction without any formation of by-products.


  • Resistance to peroxide formation improves laboratory safety
  • One-step reaction saves energy and reduces wastewater
  • More stable than tetrahydrofuran
  • Higher hydrophobicity increases yields and selectivity
  • Limited miscibility in water reduces waste stream

Formel: C₆H₁₂O
Molecular Weight: 100.16 g/mol
Schmelzpunkt: –140 °C
Dichte: 0.86 g/cm3 (20 °C)
Flammpunkt: –1 °C
MDL: MFCD08276401
CAS-Nummer: 5614-37-9
UN: 3271
ADR: 3 II

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR