Toluol, wasserfrei (max. 0,005% H₂O), SeccoSolv®, Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
Synonyme:
Anisen, Methylbenzol, Methylbenzen, Toluen, Retinnaphtha, Phenylmethan
1.08326.1001
1.08326.1001EA
0
EUR
1.08326.1001
Toluol, wasserfrei (max. 0,005% H₂O), SeccoSolv®, Sigma-Aldrich®
Toluol
Formel:
C₆H₅CH₃ Molecular Weight: 92,14 g/mol Siedepunkt: 110,6 °C (1013 hPa) Schmelzpunkt: –95 °C Dichte: 0,867 g/cm³ (20 °C) Flammpunkt: 4 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008512 CAS-Nummer: 108-88-3 EINECS: 203-625-9 UN: 1294 ADR: 3,II REACH: 05-2114615130-69 Merck Index: 13,09607 |
|
Spezifikation Testergebnisse
Solubility | 0.52 g/l (20 °C) |
Melting Point | -95 °C |
Molar Mass | 92.14 g/mol |
Boiling Point | 110.6 °C (1013 hPa) |
Vapor Pressure | 29 hPa (20 °C) |
Flash Point | 4 °C |
Refractive Index | 1.4968 (20 °C) |
Explosion Limit | 1.2 - 8 %(V) |
Density | 0.87 g/cm3 (20 °C) |
pH | (H2O) not applicable |
Ignition Point | 535 °C |
Viscosity kinematic | 0.7 mm2/s (20 °C) |
Hazard Class | 3 |
Hazard Class Description | Flammable liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...